Product Details

SNP ID
rs200031382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15475628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCACTGAAGCTGGAGGTGTACTG[G/T]CTCCAGCCTCTGTAGAAGGACAAGG
Phenotype
MIM: 608199
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PGLYRP2 PubMed Links

Gene Details

Gene
PGLYRP2
Gene Name
peptidoglycan recognition protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052890.3 1175 Missense Mutation ACA,CCA T348P NP_443122.3
XM_006722633.3 1175 Missense Mutation ACA,CCA T348P XP_006722696.1

View Full Product Details