Product Details

SNP ID
rs200241660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12133284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCTCCAGTGTGAGTCATCTTA[C/T]GATACTGAATGGAACTGAAAGAAAT
Phenotype
MIM: 194557
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF20 PubMed Links

Gene Details

Gene
ZNF20
Gene Name
zinc finger protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203250.1 1127 Missense Mutation CAT,CGT H298R NP_001190179.1
NM_021143.3 1127 Missense Mutation CAT,CGT H301R NP_066966.2
Gene
ZNF625-ZNF20
Gene Name
ZNF625-ZNF20 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details