Product Details
- SNP ID
-
rs199872237
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:15424334 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTCATTGGTGTCCTCGCCTCCCC[A/G]GGCTGCGGAGGCATCTGGAGGGCGG
- Phenotype
-
MIM: 609475
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AKAP8L
PubMed Links
Gene Details
- Gene
- AKAP8L
- Gene Name
- A-kinase anchoring protein 8 like
There are no transcripts associated with this gene.
- Gene
- WIZ
- Gene Name
- widely interspaced zinc finger motifs
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021241.2 |
3367 |
Missense Mutation |
CGG,TGG |
R692W |
NP_067064.2 |
XM_005260004.4 |
3367 |
Missense Mutation |
CGG,TGG |
R1787W |
XP_005260061.1 |
XM_005260005.4 |
3367 |
Missense Mutation |
CGG,TGG |
R1730W |
XP_005260062.1 |
XM_005260006.4 |
3367 |
Missense Mutation |
CGG,TGG |
R1654W |
XP_005260063.1 |
XM_005260007.4 |
3367 |
Missense Mutation |
CGG,TGG |
R1597W |
XP_005260064.1 |
XM_005260008.2 |
3367 |
Missense Mutation |
CGG,TGG |
R1050W |
XP_005260065.1 |
XM_005260009.2 |
3367 |
Missense Mutation |
CGG,TGG |
R866W |
XP_005260066.1 |
XM_005260010.3 |
3367 |
Missense Mutation |
CGG,TGG |
R866W |
XP_005260067.1 |
XM_005260011.2 |
3367 |
Missense Mutation |
CGG,TGG |
R860W |
XP_005260068.1 |
XM_005260012.2 |
3367 |
Missense Mutation |
CGG,TGG |
R733W |
XP_005260069.1 |
XM_006722828.3 |
3367 |
Missense Mutation |
CGG,TGG |
R1013W |
XP_006722891.1 |
XM_011528163.2 |
3367 |
Missense Mutation |
CGG,TGG |
R866W |
XP_011526465.1 |
View Full Product Details