Product Details

SNP ID
rs200290556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7477306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCACTAGCCACGGCGGGAAGC[A/G]GCCGGCCCACAGCACGCCCCGGGGC
Phenotype
MIM: 616432 MIM: 607583
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF18 PubMed Links

Gene Details

Gene
ARHGEF18
Gene Name
Rho/Rac guanine nucleotide exchange factor 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130955.1 842 Intron NP_001124427.1
NM_015318.3 842 Intron NP_056133.2
XM_005272464.4 842 Intron XP_005272521.1
XM_006722705.3 842 Intron XP_006722768.1
XM_006722706.3 842 Intron XP_006722769.1
XM_006722708.2 842 Intron XP_006722771.1
XM_011527835.2 842 Intron XP_011526137.1
XM_011527836.2 842 Intron XP_011526138.1
XM_011527837.2 842 Intron XP_011526139.1
XM_011527838.2 842 Intron XP_011526140.1
XM_011527839.2 842 Intron XP_011526141.1
XM_011527840.1 842 Intron XP_011526142.1
XM_011527841.2 842 Intron XP_011526143.1
Gene
LOC100128573
Gene Name
uncharacterized LOC100128573
There are no transcripts associated with this gene.

Gene
PEX11G
Gene Name
peroxisomal biogenesis factor 11 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270539.1 842 Intron NP_001257468.1
NM_001300881.1 842 Missense Mutation CGC,TGC R138C NP_001287810.1
NM_080662.3 842 Missense Mutation CGC,TGC R208C NP_542393.1
XM_005272506.2 842 Missense Mutation CGC,TGC R187C XP_005272563.1
XM_011528426.2 842 Intron XP_011526728.1
XM_011528427.2 842 Intron XP_011526729.1
XM_011528428.1 842 Missense Mutation CGC,TGC R138C XP_011526730.1
XM_011528429.2 842 Missense Mutation CGC,TGC R138C XP_011526731.1
XM_011528431.2 842 Missense Mutation CGC,TGC R138C XP_011526733.1
XM_011528432.1 842 Intron XP_011526734.1

View Full Product Details