Product Details

SNP ID
rs199663633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17724190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGTCCTCCTGAACCCATCAGA[C/T]AAGTCCCTGTATGATGAGGTAGGGA
Phenotype
MIM: 607573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP1S PubMed Links

Gene Details

Gene
MAP1S
Gene Name
microtubule associated protein 1S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308363.1 677 Silent Mutation GAC,GAT D69D NP_001295292.1
NM_018174.5 677 Silent Mutation GAC,GAT D95D NP_060644.4
XM_017026930.1 677 Silent Mutation GAC,GAT D95D XP_016882419.1
XM_017026931.1 677 Silent Mutation GAC,GAT D69D XP_016882420.1

View Full Product Details