Product Details

SNP ID
rs199699934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12351020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGGTTTCCCGAAGAACTGAAGG[C/T]TTTCCCACATTCCTTACAATCATAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF442 PubMed Links

Gene Details

Gene
ZNF442
Gene Name
zinc finger protein 442
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030824.2 604 Missense Mutation ACC,GCC T189A NP_110451.1
XM_006722908.3 604 Missense Mutation ACC,GCC T182A XP_006722971.1
XM_017027316.1 604 Missense Mutation ACC,GCC T197A XP_016882805.1
XM_017027317.1 604 Missense Mutation ACC,GCC T132A XP_016882806.1
XM_017027318.1 604 Missense Mutation ACC,GCC T189A XP_016882807.1
Gene
ZNF563
Gene Name
zinc finger protein 563
There are no transcripts associated with this gene.

View Full Product Details