Product Details

SNP ID
rs199879425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21405848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGGGACACAGTACGGTAGTCA[A/G]ACCCCCAGGTAGGTGAGAGTGAATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF493 PubMed Links

Gene Details

Gene
ZNF493
Gene Name
zinc finger protein 493
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076678.2 367 Intron NP_001070146.1
NM_145326.2 367 Missense Mutation AAA,AGA K83R NP_663299.2
NM_175910.6 367 Intron NP_787106.4

View Full Product Details