Product Details

SNP ID
rs199772971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6531130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCGCGTACTGCCTTGGGCCCTG[A/G]TCGCGGGGCTGCTGCTGCTGCTGCT
Phenotype
MIM: 606182
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFSF9 PubMed Links

Gene Details

Gene
TNFSF9
Gene Name
tumor necrosis factor superfamily member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003811.3 132 Missense Mutation ATC,GTC I32V NP_003802.1

View Full Product Details