Product Details

SNP ID
rs201814204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12351048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCACATTCCTTACAATCATAGCG[C/T]TTCTTTCCAGTGTGAGGTCTTTCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF442 PubMed Links

Gene Details

Gene
ZNF442
Gene Name
zinc finger protein 442
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030824.2 576 Silent Mutation AAA,AAG K179K NP_110451.1
XM_006722908.3 576 Silent Mutation AAA,AAG K172K XP_006722971.1
XM_017027316.1 576 Silent Mutation AAA,AAG K187K XP_016882805.1
XM_017027317.1 576 Silent Mutation AAA,AAG K122K XP_016882806.1
XM_017027318.1 576 Silent Mutation AAA,AAG K179K XP_016882807.1
Gene
ZNF563
Gene Name
zinc finger protein 563
There are no transcripts associated with this gene.

View Full Product Details