Product Details

SNP ID
rs200956091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12886005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGAAGTTGGTGAGGAGGAGATC[C/T]AGGTCCCAGGTGGCGTCCGCGCCCC
Phenotype
MIM: 126350 MIM: 608801 MIM: 600599
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNASE2 PubMed Links

Gene Details

Gene
DNASE2
Gene Name
deoxyribonuclease II, lysosomal
There are no transcripts associated with this gene.

Gene
GCDH
Gene Name
glutaryl-CoA dehydrogenase
There are no transcripts associated with this gene.

Gene
KLF1
Gene Name
Kruppel like factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011527642.2 298 Silent Mutation CTA,CTG L37L XP_011525944.1

View Full Product Details