Product Details

SNP ID
rs201404472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13909843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAGCTGGGCCTGCTGGTTGACCT[C/T]TCCCCAGATGGCCTGATGATCCCTG
Phenotype
MIM: 610055
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf57 PubMed Links

Gene Details

Gene
C19orf57
Gene Name
chromosome 19 open reading frame 57
There are no transcripts associated with this gene.

Gene
CC2D1A
Gene Name
coiled-coil and C2 domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017721.4 380 Silent Mutation CTC,CTT L27L NP_060191.3
XM_005259972.2 380 Silent Mutation CTC,CTT L27L XP_005260029.1
XM_005259973.2 380 Silent Mutation CTC,CTT L27L XP_005260030.1
XM_005259974.2 380 Silent Mutation CTC,CTT L27L XP_005260031.1
XM_005259975.2 380 Silent Mutation CTC,CTT L27L XP_005260032.1

View Full Product Details