Product Details
- SNP ID
-
rs200990546
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:9296190 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTTTTCTCATATGGATACTTAAG[A/G]AGGAGGGACAATTGTAGGCTTTCCC
- Phenotype
-
MIM: 609571
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF699
PubMed Links
Gene Details
- Gene
- ZNF699
- Gene Name
- zinc finger protein 699
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198535.2 |
1584 |
Missense Mutation |
TCC,TTC |
S405F |
NP_940937.1 |
View Full Product Details