Product Details

SNP ID
rs201749590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49016151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCCCCACAGTCAGAGGTGCTGC[A/G]GCGGCAGGGTCCACAGCGACAGCTG
Phenotype
MIM: 118860 MIM: 152780 MIM: 604788
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CGB3 PubMed Links

Gene Details

Gene
CGB3
Gene Name
chorionic gonadotropin beta subunit 3
There are no transcripts associated with this gene.

Gene
LHB
Gene Name
luteinizing hormone beta polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000894.2 Intron NP_000885.1
XM_011526975.1 Intron XP_011525277.1
Gene
LOC101059948
Gene Name
uncharacterized LOC101059948
There are no transcripts associated with this gene.

Gene
MIR6798
Gene Name
microRNA 6798
There are no transcripts associated with this gene.

Gene
RUVBL2
Gene Name
RuvB like AAA ATPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321190.1 Intron NP_001308119.1
NM_001321191.1 Intron NP_001308120.1
NM_006666.2 Intron NP_006657.1
XM_011526330.1 Intron XP_011524632.1

View Full Product Details