Product Details

SNP ID
rs201818743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57572789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCTCCTGTGTGAGTTCTCTGG[C/T]GTCGAACAAGAGTGGATTTGGACCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF416 PubMed Links

Gene Details

Gene
ZNF416
Gene Name
zinc finger protein 416
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017879.1 1286 Missense Mutation CAC,CGC H372R NP_060349.1
XM_005259051.3 1286 Missense Mutation CAC,CGC H300R XP_005259108.1

View Full Product Details