Product Details
- SNP ID
-
rs201996505
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12527524 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCACATTCCTGACATTTATATGGT[C/G]CATCTCCAGTGTGCTTAATCATGTG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF564
PubMed Links
Gene Details
- Gene
- ZNF564
- Gene Name
- zinc finger protein 564
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144976.3 |
794 |
Missense Mutation |
GCA,GGA |
A195G |
NP_659413.1 |
View Full Product Details