Product Details

SNP ID
rs201786850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51643933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGAGGGCTTTTCCCTCCCGGAA[A/C]CAGCTCAGTGAGGCAGGGGGGTTGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SIGLEC14 PubMed Links

Gene Details

Gene
SIGLEC14
Gene Name
sialic acid binding Ig like lectin 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098612.1 978 Missense Mutation TGG,TGT W286C NP_001092082.1
XM_017026113.1 978 Intron XP_016881602.1

View Full Product Details