Product Details

SNP ID
rs202242420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15238869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCAGCCACCGCAGCTGCTTGCTG[A/T]TGCTGCTGCTGCTGTTGCTCCTGGC
Phenotype
MIM: 608749
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BRD4 PubMed Links

Gene Details

Gene
BRD4
Gene Name
bromodomain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014299.2 4116 Intron NP_055114.1
NM_058243.2 4116 Missense Mutation CAA,CAT Q1298H NP_490597.1
XM_011527854.1 4116 Missense Mutation CAA,CAT Q1298H XP_011526156.1
XM_011527855.1 4116 Intron XP_011526157.1
XM_011527856.2 4116 Intron XP_011526158.1
Gene
EPHX3
Gene Name
epoxide hydrolase 3
There are no transcripts associated with this gene.

View Full Product Details