Product Details

SNP ID
rs200584353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19630000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGCTTCCTCAGGCTGGACGTCC[A/G]GGCAGCAGGTGGCCCTGGGCACAGC
Phenotype
MIM: 609694 MIM: 605110
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GMIP PubMed Links

Gene Details

Gene
GMIP
Gene Name
GEM interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288998.1 2927 Missense Mutation CCG,CTG P930L NP_001275927.1
NM_001288999.1 2927 Missense Mutation CCG,CTG P933L NP_001275928.1
NM_016573.3 2927 Missense Mutation CCG,CTG P959L NP_057657.2
XM_005259927.2 2927 Missense Mutation CCG,CTG P956L XP_005259984.1
XM_011528048.2 2927 Missense Mutation CCG,CTG P838L XP_011526350.1
XM_017026861.1 2927 UTR 3 XP_016882350.1
XM_017026862.1 2927 Intron XP_016882351.1
Gene
LPAR2
Gene Name
lysophosphatidic acid receptor 2
There are no transcripts associated with this gene.

View Full Product Details