Product Details

SNP ID
rs201104603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58453780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACACAGCGCAGAGGGCCCTGTAC[C/T]GCCACGTGATGCTGGAAAACTTCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF324B PubMed Links

Gene Details

Gene
ZNF324B
Gene Name
zinc finger protein 324B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207395.2 210 Missense Mutation CGC,TGC R27C NP_997278.2
XM_005258915.4 210 Missense Mutation CGC,TGC R27C XP_005258972.1
XM_005258918.4 210 Missense Mutation CGC,TGC R27C XP_005258975.1

View Full Product Details