Product Details

SNP ID
rs201209520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54631500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTGACTCCTGATTTCCTTCCAG[A/G]GCACCTCCCCAAGCCCACCCTCTGG
Phenotype
MIM: 604811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LILRB1 PubMed Links

Gene Details

Gene
LILRB1
Gene Name
leukocyte immunoglobulin like receptor B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081637.2 342 Intron NP_001075106.2
NM_001081638.3 342 Missense Mutation GAG,GGG E24G NP_001075107.2
NM_001081639.3 342 Missense Mutation GAG,GGG E24G NP_001075108.2
NM_001278398.2 342 Intron NP_001265327.2
NM_001278399.2 342 Missense Mutation GAG,GGG E24G NP_001265328.2
NM_006669.6 342 Missense Mutation GAG,GGG E24G NP_006660.4
XM_011526331.2 342 Intron XP_011524633.1
XM_011526332.2 342 Intron XP_011524634.1
XM_011526335.2 342 Intron XP_011524637.1
XM_011526336.2 342 Intron XP_011524638.1
XM_011526338.2 342 Intron XP_011524640.1
XM_017026182.1 342 Intron XP_016881671.1
XM_017026183.1 342 Intron XP_016881672.1
XM_017026184.1 342 Intron XP_016881673.1
XM_017026185.1 342 Intron XP_016881674.1
XM_017026186.1 342 Intron XP_016881675.1
XM_017026187.1 342 Missense Mutation GAG,GGG E41G XP_016881676.1
XM_017026188.1 342 Missense Mutation GAG,GGG E41G XP_016881677.1
XM_017026189.1 342 Missense Mutation GAG,GGG E41G XP_016881678.1
XM_017026190.1 342 Missense Mutation GAG,GGG E41G XP_016881679.1
XM_017026191.1 342 Intron XP_016881680.1
XM_017026192.1 342 Intron XP_016881681.1

View Full Product Details