Product Details

SNP ID
rs201694256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44476702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAGTATGAGTTCTCTGATGTAC[A/G]ACAAGAACATAACTCTGGCTGAAGG
Phenotype
MIM: 606740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF180 PubMed Links

Gene Details

Gene
ZNF180
Gene Name
zinc finger protein 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278508.2 1940 Silent Mutation GTC,GTT V568V NP_001265437.2
NM_001278509.2 1940 Silent Mutation GTC,GTT V566V NP_001265438.2
NM_001288759.2 1940 Silent Mutation GTC,GTT V592V NP_001275688.2
NM_001288760.2 1940 Silent Mutation GTC,GTT V250V NP_001275689.1
NM_001288761.2 1940 Silent Mutation GTC,GTT V250V NP_001275690.1
NM_001288762.2 1940 Silent Mutation GTC,GTT V250V NP_001275691.1
NM_001291633.1 1940 Silent Mutation GTC,GTT V566V NP_001278562.1
NM_013256.5 1940 Silent Mutation GTC,GTT V593V NP_037388.3
XM_011527280.2 1940 Silent Mutation GTC,GTT V635V XP_011525582.1
XM_017027250.1 1940 Silent Mutation GTC,GTT V250V XP_016882739.1
XM_017027251.1 1940 Silent Mutation GTC,GTT V250V XP_016882740.1
XM_017027252.1 1940 Silent Mutation GTC,GTT V250V XP_016882741.1
XM_017027253.1 1940 Silent Mutation GTC,GTT V250V XP_016882742.1
XM_017027254.1 1940 Silent Mutation GTC,GTT V250V XP_016882743.1
XM_017027255.1 1940 Silent Mutation GTC,GTT V250V XP_016882744.1
XM_017027256.1 1940 Silent Mutation GTC,GTT V250V XP_016882745.1

View Full Product Details