Product Details

SNP ID
rs202008767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5693307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACAGGGACACTCACCGCAATGGT[C/T]TTCTCCTTGATGCCACCAACAGGCA
Phenotype
MIM: 605490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSD11B1L PubMed Links

Gene Details

Gene
HSD11B1L
Gene Name
hydroxysteroid 11-beta dehydrogenase 1 like
There are no transcripts associated with this gene.

Gene
LONP1
Gene Name
lon peptidase 1, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276479.1 2535 Silent Mutation AAA,AAG K834K NP_001263408.1
NM_001276480.1 2535 Silent Mutation AAA,AAG K702K NP_001263409.1
NM_004793.3 2535 Silent Mutation AAA,AAG K898K NP_004784.2
XM_011528441.2 2535 Intron XP_011526743.1
Gene
RPL36
Gene Name
ribosomal protein L36
There are no transcripts associated with this gene.

View Full Product Details