Product Details

SNP ID
rs201211116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14028346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCGAGCAGTCATCTTCACCTGC[G/T]GGGGCTCCCGGTGGAGACGATCAGA
Phenotype
MIM: 605350 MIM: 606855
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL27RA PubMed Links

Gene Details

Gene
IL27RA
Gene Name
interleukin 27 receptor subunit alpha
There are no transcripts associated with this gene.

Gene
RLN3
Gene Name
relaxin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311197.1 199 Missense Mutation GGG,TGG G48W NP_001298126.1
NM_080864.3 199 Missense Mutation GGG,TGG G48W NP_543140.1

View Full Product Details