Product Details

SNP ID
rs201490930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:685778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGGCCGGGCTGGGGACTGCTCC[A/G]CCGAACCACGTCCCAGATCCAGGCC
Phenotype
MIM: 605343
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FSTL3 PubMed Links

Gene Details

Gene
FSTL3
Gene Name
follistatin like 3
There are no transcripts associated with this gene.

Gene
PRSS57
Gene Name
protease, serine 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308209.1 818 Missense Mutation CGG,TGG R263W NP_001295138.1
NM_214710.4 818 Missense Mutation CGG,TGG R264W NP_999875.1

View Full Product Details