Product Details

SNP ID
rs201076390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57506403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGGCAGCTGCTGAGCAGAGTG[C/T]TTCTGTAGAAGTGCCCAGTTCAAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF773 PubMed Links

Gene Details

Gene
ZNF773
Gene Name
zinc finger protein 773
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304334.1 532 Missense Mutation GCT,GTT A102V NP_001291263.1
NM_001304335.1 532 Intron NP_001291264.1
NM_001304336.1 532 Missense Mutation CTT,TTT L101F NP_001291265.1
NM_001304337.1 532 Intron NP_001291266.1
NM_198542.2 532 Missense Mutation GCT,GTT A103V NP_940944.1

View Full Product Details