Product Details

SNP ID
rs202013937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12133130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCTTTTCATGCCTTTGAAGGTC[C/T]GAGGTACATCTGAAGGCTTTACCAC
Phenotype
MIM: 194557
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF20 PubMed Links

Gene Details

Gene
ZNF20
Gene Name
zinc finger protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203250.1 1281 Silent Mutation TCA,TCG S349S NP_001190179.1
NM_021143.3 1281 Silent Mutation TCA,TCG S352S NP_066966.2
Gene
ZNF625-ZNF20
Gene Name
ZNF625-ZNF20 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details