Product Details

SNP ID
rs201081309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41220654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCAGGCACGCAGGCTGAAGAAA[G/T]TCCCTTCGTGGGCAACCCAGGGAAT
Phenotype
MIM: 109135
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AXL PubMed Links

Gene Details

Gene
AXL
Gene Name
AXL receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278599.1 298 Intron NP_001265528.1
NM_001699.5 298 Missense Mutation AGT,ATT S35I NP_001690.2
NM_021913.4 298 Missense Mutation AGT,ATT S35I NP_068713.2

View Full Product Details