Product Details

SNP ID
rs201806494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35777863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCCCCTTCCCGCGGCTGGCTGA[C/G]TGCGCCCATTTCCACTACGAGAACG
Phenotype
MIM: 614902
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP33 PubMed Links

Gene Details

Gene
ARHGAP33
Gene Name
Rho GTPase activating protein 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172630.1 1684 UTR 5 NP_001166101.1
NM_052948.3 1684 Missense Mutation GAC,GAG D48E NP_443180.2
XM_005258488.1 1684 Missense Mutation GAC,GAG D48E XP_005258545.1
XM_006722999.1 1684 Missense Mutation GAC,GAG D48E XP_006723062.1
XM_006723003.2 1684 Intron XP_006723066.1
XM_011526416.1 1684 UTR 5 XP_011524718.1
XM_011526417.1 1684 UTR 5 XP_011524719.1
XM_011526418.1 1684 UTR 5 XP_011524720.1
XM_011526419.1 1684 UTR 5 XP_011524721.1
XM_011526420.1 1684 UTR 5 XP_011524722.1
XM_011526421.2 1684 Missense Mutation GAC,GAG D85E XP_011524723.1
XM_017026238.1 1684 Missense Mutation GAC,GAG D85E XP_016881727.1
Gene
PROSER3
Gene Name
proline and serine rich 3
There are no transcripts associated with this gene.

View Full Product Details