Product Details

SNP ID
rs202226528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34404769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCGTTTCACCGTCTGCTGCAC[G/T]TGTTCGCGTGCGCCTGCCCCGGCTG
Phenotype
MIM: 172400 MIM: 615661
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPI PubMed Links

Gene Details

Gene
GPI
Gene Name
glucose-6-phosphate isomerase
There are no transcripts associated with this gene.

Gene
PDCD2L
Gene Name
programmed cell death 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032346.1 262 Missense Mutation GTG,TTG V77L NP_115722.1

View Full Product Details