Product Details

SNP ID
rs201051792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40379679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGGGGTGGGAGCCCCTGAGCCC[A/G]TGGGCCCCTGACGCTCTCCAGGGTG
Phenotype
MIM: 611712 MIM: 615698
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIPK4 PubMed Links

Gene Details

Gene
HIPK4
Gene Name
homeodomain interacting protein kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144685.4 2062 Missense Mutation CGG,TGG R587W NP_653286.2
XM_006723036.2 2062 Missense Mutation CGG,TGG R590W XP_006723099.1
Gene
MIR6796
Gene Name
microRNA 6796
There are no transcripts associated with this gene.

Gene
PLD3
Gene Name
phospholipase D family member 3
There are no transcripts associated with this gene.

View Full Product Details