Product Details

SNP ID
rs201533714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40380433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCTCCTTCCTCCCAGCTGCTGC[C/T]CCGGCAGGGCCTGTCATCCTCAGGC
Phenotype
MIM: 611712 MIM: 615698
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIPK4 PubMed Links

Gene Details

Gene
HIPK4
Gene Name
homeodomain interacting protein kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144685.4 1852 Missense Mutation AGC,GGC S520G NP_653286.2
XM_006723036.2 1852 Missense Mutation AGC,GGC S520G XP_006723099.1
Gene
PLD3
Gene Name
phospholipase D family member 3
There are no transcripts associated with this gene.

View Full Product Details