Product Details

SNP ID
rs201224544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54631506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCTGATTTCCTTCCAGGGCACC[A/T]CCCCAAGCCCACCCTCTGGGCTGAA
Phenotype
MIM: 604811
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LILRB1 PubMed Links

Gene Details

Gene
LILRB1
Gene Name
leukocyte immunoglobulin like receptor B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081637.2 348 Intron NP_001075106.2
NM_001081638.3 348 Missense Mutation CAC,CTC H26L NP_001075107.2
NM_001081639.3 348 Missense Mutation CAC,CTC H26L NP_001075108.2
NM_001278398.2 348 Intron NP_001265327.2
NM_001278399.2 348 Missense Mutation CAC,CTC H26L NP_001265328.2
NM_006669.6 348 Missense Mutation CAC,CTC H26L NP_006660.4
XM_011526331.2 348 Intron XP_011524633.1
XM_011526332.2 348 Intron XP_011524634.1
XM_011526335.2 348 Intron XP_011524637.1
XM_011526336.2 348 Intron XP_011524638.1
XM_011526338.2 348 Intron XP_011524640.1
XM_017026182.1 348 Intron XP_016881671.1
XM_017026183.1 348 Intron XP_016881672.1
XM_017026184.1 348 Intron XP_016881673.1
XM_017026185.1 348 Intron XP_016881674.1
XM_017026186.1 348 Intron XP_016881675.1
XM_017026187.1 348 Missense Mutation CAC,CTC H43L XP_016881676.1
XM_017026188.1 348 Missense Mutation CAC,CTC H43L XP_016881677.1
XM_017026189.1 348 Missense Mutation CAC,CTC H43L XP_016881678.1
XM_017026190.1 348 Missense Mutation CAC,CTC H43L XP_016881679.1
XM_017026191.1 348 Intron XP_016881680.1
XM_017026192.1 348 Intron XP_016881681.1

View Full Product Details