Product Details
- SNP ID
-
rs201124053
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:159588139 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGTTTGTATTTCCTAGAGAATCT[C/G]TTACTGATCATGTAAACTTGATCAC
- Phenotype
-
MIM: 104770
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
APCS
PubMed Links
Gene Details
- Gene
- APCS
- Gene Name
- amyloid P component, serum
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001639.3 |
199 |
Missense Mutation |
CTT,GTT |
L35V |
NP_001630.1 |
View Full Product Details