Product Details

SNP ID
rs201257450
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:150267462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGAAGACACCACTGATGATACC[A/G]AAGGAGAGACCAGAAACTGGGGGAA
Phenotype
MIM: 607629 MIM: 604832
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APH1A PubMed Links

Gene Details

Gene
APH1A
Gene Name
aph-1 homolog A, gamma-secretase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077628.2 774 Silent Mutation TTC,TTT F125F NP_001071096.1
NM_001243771.1 774 Silent Mutation TTC,TTT F68F NP_001230700.1
NM_001243772.1 774 Silent Mutation TTC,TTT F55F NP_001230701.1
NM_016022.3 774 Silent Mutation TTC,TTT F125F NP_057106.2
XM_017001417.1 774 Silent Mutation TTC,TTT F68F XP_016856906.1
Gene
C1orf54
Gene Name
chromosome 1 open reading frame 54
There are no transcripts associated with this gene.

Gene
CA14
Gene Name
carbonic anhydrase 14
There are no transcripts associated with this gene.

View Full Product Details