Product Details

SNP ID
rs201454895
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:178514049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCAAGAACTACAAGGCAGTTT[G/T]CCTGGAATTGAAGCCAGAGCCGACC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TEX35 PubMed Links

Gene Details

Gene
TEX35
Gene Name
testis expressed 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170722.1 174 Missense Mutation TGC,TTC C29F NP_001164193.1
NM_001170723.1 174 Missense Mutation TGC,TTC C21F NP_001164194.1
NM_001170724.1 174 Missense Mutation TGC,TTC C21F NP_001164195.1
NM_032126.4 174 Missense Mutation TGC,TTC C21F NP_115502.2

View Full Product Details