Product Details

SNP ID
rs201259305
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:247532009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGATGCAATTGTTCCCACACAGC[A/G]GTAGGAGCATGGTGAGCGTGGAGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GCSAML PubMed Links

Gene Details

Gene
GCSAML
Gene Name
germinal center associated signaling and motility like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281834.1 865 Intron NP_001268763.1
NM_001281835.1 865 Intron NP_001268764.1
NM_001281836.1 865 Intron NP_001268765.1
NM_001281837.1 865 Intron NP_001268766.1
NM_001281838.1 865 Intron NP_001268767.1
NM_001281853.1 865 Intron NP_001268782.1
NM_145278.4 865 Intron NP_660321.1
XM_017000399.1 865 Intron XP_016855888.1
Gene
GCSAML-AS1
Gene Name
GCSAML antisense RNA 1
There are no transcripts associated with this gene.

Gene
OR2C3
Gene Name
olfactory receptor family 2 subfamily C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198074.4 865 Missense Mutation NP_932340.3

View Full Product Details