Product Details
- SNP ID
-
rs201337220
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:15617778 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAACTTCCGCGCGCTGTGCGAGCT[C/G]GAGTCTGGCATCCCCGCAGCCGAGA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DDI2
PubMed Links
Gene Details
- Gene
- DDI2
- Gene Name
- DNA damage inducible 1 homolog 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032341.4 |
321 |
Silent Mutation |
CTC,CTG |
L36L |
NP_115717.3 |
View Full Product Details