Product Details

SNP ID
rs201734412
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:43935670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTGATAGAGATGGAACTTGGAC[C/T]TGGAGGCCTCTCCACGCTGTCCCAC
Phenotype
MIM: 603886 MIM: 606494
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARTN PubMed Links

Gene Details

Gene
ARTN
Gene Name
artemin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136215.1 796 Missense Mutation CCT,CTT P5L NP_001129687.1
NM_057090.2 796 Missense Mutation CCT,CTT P5L NP_476431.2
NM_057091.2 796 Missense Mutation CCT,CTT P5L NP_476432.2
Gene
ST3GAL3
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
There are no transcripts associated with this gene.

View Full Product Details