Product Details

SNP ID
rs201770232
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:158398914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCAGTGCAGGCTAACTTGATAA[C/G]AGGTGCCATGTCACAGAAATAGTGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR10T2 PubMed Links

Gene Details

Gene
OR10T2
Gene Name
olfactory receptor family 10 subfamily T member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004475.1 553 Missense Mutation CTT,GTT L185V NP_001004475.1

View Full Product Details