Product Details
- SNP ID
-
rs200983551
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:152154875 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTCATGGGTTTGTCTGTCTCGTC[C/T]CTGATGGTTCTGCTCATCTTTATGG
- Phenotype
-
MIM: 613259
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RPTN
PubMed Links
Gene Details
- Gene
- RPTN
- Gene Name
- repetin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001122965.1 |
2289 |
Missense Mutation |
AGA,GGA |
R742G |
NP_001116437.1 |
View Full Product Details