Product Details
- SNP ID
-
rs201262383
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:19814802 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGCCCCACCACCGCCTCATGGTC[A/G]CGCAGGCTGCAGATGAGGCCCCCGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNF186
PubMed Links
Gene Details
- Gene
- RNF186
- Gene Name
- ring finger protein 186
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019062.1 |
477 |
Silent Mutation |
CGC,CGT |
R100R |
NP_061935.1 |
View Full Product Details