Product Details

SNP ID
rs201116540
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:68431066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACACAACAATTGCTTTCATTACC[A/G]TCATCTTCTTCCAAGGCATCTGGGT
Phenotype
MIM: 180069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPE65 PubMed Links

Gene Details

Gene
RPE65
Gene Name
RPE65, retinoid isomerohydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000329.2 1715 Silent Mutation GAC,GAT D483D NP_000320.1
XM_017002027.1 1715 Silent Mutation GAC,GAT D391D XP_016857516.1

View Full Product Details