Product Details

SNP ID
rs201698271
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:167055052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGTAGATGATGATGCCAATGA[C/T]AATGAGGGCTGCAACCACGCCCACC
Phenotype
MIM: 602171
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPA33 PubMed Links

Gene Details

Gene
GPA33
Gene Name
glycoprotein A33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005814.2 923 Missense Mutation ATC,GTC I251V NP_005805.1
XM_017000005.1 923 Missense Mutation ATC,GTC I143V XP_016855494.1

View Full Product Details