Product Details
- SNP ID
-
rs200972450
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:46558590 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGAGCATTTCAGAGTGCTGTGGG[C/T]GGGCTCCTGTCTTCACCACGGCCTG
- Phenotype
-
MIM: 611455
MIM: 606724
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KNCN
PubMed Links
Gene Details
- Gene
- KNCN
- Gene Name
- kinocilin
There are no transcripts associated with this gene.
- Gene
- MKNK1
- Gene Name
- MAP kinase interacting serine/threonine kinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001135553.2 |
1474 |
Silent Mutation |
CCA,CCG |
P420P |
NP_001129025.1 |
NM_003684.5 |
1474 |
Silent Mutation |
CCA,CCG |
P461P |
NP_003675.2 |
NM_198973.3 |
1474 |
UTR 3 |
|
|
NP_945324.1 |
XM_006711000.1 |
1474 |
Silent Mutation |
CCA,CCG |
P420P |
XP_006711063.1 |
XM_006711001.2 |
1474 |
Silent Mutation |
CCA,CCG |
P414P |
XP_006711064.1 |
XM_006711002.2 |
1474 |
Silent Mutation |
CCA,CCG |
P414P |
XP_006711065.1 |
XM_006711003.1 |
1474 |
Silent Mutation |
CCA,CCG |
P414P |
XP_006711066.1 |
XM_006711006.3 |
1474 |
Silent Mutation |
CCA,CCG |
P365P |
XP_006711069.1 |
XM_006711007.1 |
1474 |
Silent Mutation |
CCA,CCG |
P365P |
XP_006711070.1 |
XM_006711009.3 |
1474 |
Silent Mutation |
CCA,CCG |
P365P |
XP_006711072.1 |
XM_006711013.1 |
1474 |
Silent Mutation |
CCA,CCG |
P325P |
XP_006711076.1 |
XM_006711014.1 |
1474 |
Silent Mutation |
CCA,CCG |
P325P |
XP_006711077.1 |
XM_006711015.1 |
1474 |
Silent Mutation |
CCA,CCG |
P325P |
XP_006711078.1 |
XM_017002652.1 |
1474 |
Silent Mutation |
CCA,CCG |
P423P |
XP_016858141.1 |
XM_017002653.1 |
1474 |
Silent Mutation |
CCA,CCG |
P423P |
XP_016858142.1 |
XM_017002654.1 |
1474 |
Silent Mutation |
CCA,CCG |
P423P |
XP_016858143.1 |
XM_017002655.1 |
1474 |
Silent Mutation |
CCA,CCG |
P408P |
XP_016858144.1 |
XM_017002656.1 |
1474 |
Silent Mutation |
CCA,CCG |
P408P |
XP_016858145.1 |
XM_017002657.1 |
1474 |
Silent Mutation |
CCA,CCG |
P365P |
XP_016858146.1 |
XM_017002658.1 |
1474 |
UTR 3 |
|
|
XP_016858147.1 |
XM_017002659.1 |
1474 |
UTR 3 |
|
|
XP_016858148.1 |
XM_017002660.1 |
1474 |
UTR 3 |
|
|
XP_016858149.1 |
XM_017002661.1 |
1474 |
UTR 3 |
|
|
XP_016858150.1 |
XM_017002662.1 |
1474 |
UTR 3 |
|
|
XP_016858151.1 |
XM_017002663.1 |
1474 |
UTR 3 |
|
|
XP_016858152.1 |
XM_017002664.1 |
1474 |
Silent Mutation |
CCA,CCG |
P325P |
XP_016858153.1 |
XM_017002665.1 |
1474 |
UTR 3 |
|
|
XP_016858154.1 |
XM_017002666.1 |
1474 |
UTR 3 |
|
|
XP_016858155.1 |
XM_017002667.1 |
1474 |
UTR 3 |
|
|
XP_016858156.1 |
XM_017002668.1 |
1474 |
Intron |
|
|
XP_016858157.1 |
- Gene
- MKNK1-AS1
- Gene Name
- MKNK1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details