Product Details

SNP ID
rs201712832
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:234607563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCCTCCTGGTAGTGGAGTGAAC[C/G]TGGTTGGCATCTTTTGAGGCATGAC
Phenotype
MIM: 615332
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IRF2BP2 PubMed Links

Gene Details

Gene
IRF2BP2
Gene Name
interferon regulatory factor 2 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077397.1 1369 Missense Mutation CAC,CAG H430Q NP_001070865.1
NM_182972.2 1369 Missense Mutation CAC,CAG H446Q NP_892017.2

View Full Product Details