Product Details

SNP ID
rs200999217
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:153662513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTCATAAGCCTTTTCTGAAGG[C/T]GTTACTATCACTCCATCCCAGGTAG
Phenotype
MIM: 603181 MIM: 607007
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ILF2 PubMed Links

Gene Details

Gene
ILF2
Gene Name
interleukin enhancer binding factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267809.1 1162 Silent Mutation ACA,ACG T314T NP_001254738.1
NM_004515.3 1162 Silent Mutation ACA,ACG T352T NP_004506.2
Gene
SNAPIN
Gene Name
SNAP associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012437.5 1162 Intron NP_036569.1

View Full Product Details