Product Details

SNP ID
rs201726959
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:159587927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGCCCTAGGCCAGGAATATGAA[A/C]AAGCCGCTGCTTTGGATCTCTGTCC
Phenotype
MIM: 104770
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APCS PubMed Links

Gene Details

Gene
APCS
Gene Name
amyloid P component, serum
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001639.3 102 Missense Mutation AAA,AAC K2N NP_001630.1

View Full Product Details