Product Details

SNP ID
rs201038287
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:113073426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAAATGGCGCCGGCGCCCCTAG[C/G]CGTCCCGGAGGAGCAGTTGCTGGGG
Phenotype
MIM: 608869
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100996251 PubMed Links

Gene Details

Gene
LOC100996251
Gene Name
uncharacterized LOC100996251
There are no transcripts associated with this gene.

Gene
LRIG2
Gene Name
leucine rich repeats and immunoglobulin like domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001312686.1 257 UTR 5 NP_001299615.1
NM_014813.2 257 Missense Mutation GCC,GGC A7G NP_055628.1
XM_005271369.2 257 Missense Mutation GCC,GGC A7G XP_005271426.1
XM_017002952.1 257 Intron XP_016858441.1

View Full Product Details