Product Details
- SNP ID
-
rs201344469
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:219179215 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATCAAGATTTAACATTCCAACAC[A/T]TAAAAATTATTTATCCAACAGCTCC
- Phenotype
-
MIM: 616548
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LYPLAL1
PubMed Links
Gene Details
- Gene
- LYPLAL1
- Gene Name
- lysophospholipase like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001300769.1 |
484 |
Missense Mutation |
ATA,TTA |
I54L |
NP_001287698.1 |
NM_001300770.1 |
484 |
Missense Mutation |
ATA,TTA |
I54L |
NP_001287699.1 |
NM_001300771.1 |
484 |
Missense Mutation |
ATA,TTA |
I16L |
NP_001287700.1 |
NM_001300772.1 |
484 |
Missense Mutation |
ATA,TTA |
I16L |
NP_001287701.1 |
NM_138794.4 |
484 |
Missense Mutation |
ATA,TTA |
I54L |
NP_620149.2 |
XM_006711162.3 |
484 |
Missense Mutation |
ATA,TTA |
I54L |
XP_006711225.1 |
XM_011509182.2 |
484 |
Missense Mutation |
ATA,TTA |
I54L |
XP_011507484.1 |
XM_011509183.2 |
484 |
Missense Mutation |
ATA,TTA |
I16L |
XP_011507485.1 |
XM_017000271.1 |
484 |
Missense Mutation |
ATA,TTA |
I54L |
XP_016855760.1 |
XM_017000272.1 |
484 |
Missense Mutation |
ATA,TTA |
I16L |
XP_016855761.1 |
- Gene
- LYPLAL1-AS1
- Gene Name
- LYPLAL1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details