Product Details
- SNP ID
-
rs201228241
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:1084503 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACGGCTTCGCCAGGCTGCAGGGCC[A/G]GAGCTGTGGGGGAGAAAGCGGAGAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf159
PubMed Links
Gene Details
- Gene
- C1orf159
- Gene Name
- chromosome 1 open reading frame 159
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017891.4 |
847 |
Missense Mutation |
CCG,CTG |
P150L |
NP_060361.4 |
XM_005244765.2 |
847 |
Missense Mutation |
CCG,CTG |
P150L |
XP_005244822.1 |
XM_006710717.2 |
847 |
Missense Mutation |
CCG,CTG |
P150L |
XP_006710780.1 |
XM_006710718.2 |
847 |
Missense Mutation |
CCG,CTG |
P150L |
XP_006710781.1 |
XM_011541651.2 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_011539953.1 |
XM_011541652.2 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_011539954.1 |
XM_011541653.2 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_011539955.1 |
XM_011541655.2 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_011539957.1 |
XM_011541656.2 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_011539958.1 |
XM_011541658.2 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_011539960.1 |
XM_011541659.2 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_011539961.1 |
XM_011541660.2 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_011539962.1 |
XM_011541661.2 |
847 |
Missense Mutation |
CCG,CTG |
P186L |
XP_011539963.1 |
XM_011541662.2 |
847 |
Missense Mutation |
CCG,CTG |
P186L |
XP_011539964.1 |
XM_011541663.2 |
847 |
Missense Mutation |
CCG,CTG |
P186L |
XP_011539965.1 |
XM_011541665.2 |
847 |
Missense Mutation |
CCG,CTG |
P150L |
XP_011539967.1 |
XM_017001572.1 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_016857061.1 |
XM_017001573.1 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_016857062.1 |
XM_017001574.1 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_016857063.1 |
XM_017001575.1 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_016857064.1 |
XM_017001576.1 |
847 |
Missense Mutation |
CCG,CTG |
P186L |
XP_016857065.1 |
XM_017001577.1 |
847 |
Missense Mutation |
CCG,CTG |
P150L |
XP_016857066.1 |
XM_017001578.1 |
847 |
Missense Mutation |
CCG,CTG |
P150L |
XP_016857067.1 |
XM_017001579.1 |
847 |
Missense Mutation |
CCG,CTG |
P150L |
XP_016857068.1 |
XM_017001580.1 |
847 |
Missense Mutation |
CCG,CTG |
P218L |
XP_016857069.1 |
View Full Product Details